SY3H304528BE Synthonix Corporation

3-Bromo-5-isopropylpyridine

  • Cas Number : 1209459-74-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/32128.html

Safety Information

MSDS https://synthonix.com/32128.html
Availability details for
Quantity
  • Availability
  • Add to Cart