SY3H304528BF Synthonix Corporation

6-Bromopyrazine-2-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 1196151-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/32134.html

Safety Information

MSDS https://synthonix.com/32134.html
Availability details for
Quantity
  • Availability
  • Add to Cart