SY3H304528C0 Synthonix Corporation

5-Bromo-1-chloro-6-methylisoquinoline

  • MDL number No MDL found
  • Cas Number : 1245647-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/32140.html

Safety Information

MSDS https://synthonix.com/32140.html
Availability details for
Quantity
  • Availability
  • Add to Cart