SY3H304528D6 Synthonix Corporation

4-Bromo-2-chloro-5-methylpyridine

  • MDL number No MDL found
  • Cas Number : 867279-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/9888.html

Safety Information

MSDS https://synthonix.com/9888.html
Availability details for
Quantity
  • Availability
  • Add to Cart