SY3H304528D8 Synthonix Corporation

2-Chloro-3-methoxy-5-nitropyridine

  • MDL number No MDL found
  • Cas Number : 75711-00-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/25245.html

Safety Information

MSDS https://synthonix.com/25245.html
Availability details for
Quantity
  • Availability
  • Add to Cart