SY3H304528DB Synthonix Corporation

5-(Chloromethyl)nicotinonitrile

  • Cas Number : 562074-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/32023.html

Safety Information

MSDS https://synthonix.com/32023.html
Availability details for
Quantity
  • Availability
  • Add to Cart