SY3H304528E3 Synthonix Corporation

2-Chlorothiazolo[4,5-b]pyridine

  • Cas Number : 152170-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/32167.html

Safety Information

MSDS https://synthonix.com/32167.html
Availability details for
Quantity
  • Availability
  • Add to Cart