SY3H304528E6 Synthonix Corporation

3-Chloro-5-nitroaniline

  • Cas Number : 5344-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/32215.html

Safety Information

MSDS https://synthonix.com/32215.html
Availability details for
Quantity
  • Availability
  • Add to Cart