SY3H304528ED Synthonix Corporation

1-[3,3-Dimethylbutynyl]-1,2-benziodoxol-3(1H)-one

  • MDL number MFCD31735561
  • Cas Number : 135226-09-4
  • Molecular Weight : 328.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/2772.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/2772.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart