SY3H304528F5 Synthonix Corporation

3,5-Dibromo-6-methylpyrazin-2-amine

  • Cas Number : 74290-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/32051.html

Safety Information

MSDS https://synthonix.com/32051.html
Availability details for
Quantity
  • Availability
  • Add to Cart