SY3H304528FF Synthonix Corporation

(4,6-Dichloropyrimidin-2-yl)methanol

  • Cas Number : 1244949-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/32146.html

Safety Information

MSDS https://synthonix.com/32146.html
Availability details for
Quantity
  • Availability
  • Add to Cart