SY3H30452902 Synthonix Corporation

2,5-Dichloro-4,6-dimethylnicotinic acid

  • Cas Number : 142266-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/32171.html

Safety Information

MSDS https://synthonix.com/32171.html
Availability details for
Quantity
  • Availability
  • Add to Cart