SY3H30452904 Synthonix Corporation

2,3-Dichloro-5-fluoropyridine

  • MDL number No MDL found
  • Cas Number : 185985-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/32190.html

Safety Information

MSDS https://synthonix.com/32190.html
Availability details for
Quantity
  • Availability
  • Add to Cart