SY3H30452906 Synthonix Corporation

2,5-Dichloroquinoxaline

  • Cas Number : 55687-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/32209.html

Safety Information

MSDS https://synthonix.com/32209.html
Availability details for
Quantity
  • Availability
  • Add to Cart