SY3H3045290A Synthonix Corporation

(1,1-Dimethylethyl)[(6-iodohexyl)oxy]dimethylsilane

  • MDL number No MDL found
  • Cas Number : 103483-32-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/8491.html

Safety Information

MSDS https://synthonix.com/8491.html
Availability details for
Quantity
  • Availability
  • Add to Cart