SY3H3045290B Synthonix Corporation

2-Ethoxy-4,6-dichloropyrimidine

  • MDL number MFCD02262171
  • Cas Number : 40758-65-4
  • Molecular Weight : 193.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/23564.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/23564.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart