SY3H30452915 Synthonix Corporation

Ethyl 5-nitro-1H-imidazole-2-carboxylate

  • Cas Number : 865998-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/32233.html

Safety Information

MSDS https://synthonix.com/32233.html
Availability details for
Quantity
  • Availability
  • Add to Cart