SY3H30452922 Synthonix Corporation

2-Fluoro-9H-purin-6-amine

  • Cas Number : 700-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/9539.html

Safety Information

MSDS https://synthonix.com/9539.html
Availability details for
Quantity
  • Availability
  • Add to Cart