SY3H30452928 Synthonix Corporation

N-Hydroxy-4-nitrobenzimidoyl chloride

  • MDL number No MDL found
  • Cas Number : 1011-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/32150.html

Safety Information

MSDS https://synthonix.com/32150.html
Availability details for
Quantity
  • Availability
  • Add to Cart