SY3H30452929 Synthonix Corporation

tert-Butyl (5-(2-hydroxyethyl)pyridin-2-yl)carbamate

  • Cas Number : 1260897-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/32152.html

Safety Information

MSDS https://synthonix.com/32152.html
Availability details for
Quantity
  • Availability
  • Add to Cart