SY3H30452931 Synthonix Corporation

Isoquinolin-3-ylmethanamine

  • MDL number No MDL found
  • Cas Number : 132833-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/32057.html

Safety Information

MSDS https://synthonix.com/32057.html
Availability details for
Quantity
  • Availability
  • Add to Cart