SY3H30452939 Synthonix Corporation

2-Methyl-1,3,4-oxadiazole

  • MDL number No MDL found
  • Cas Number : 3451-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/32022.html

Safety Information

MSDS https://synthonix.com/32022.html
Availability details for
Quantity
  • Availability
  • Add to Cart