SY3H3045293B Synthonix Corporation

Methyl cinnoline-6-carboxylate

  • MDL number No MDL found
  • Cas Number : 318276-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/32046.html

Safety Information

MSDS https://synthonix.com/32046.html
Availability details for
Quantity
  • Availability
  • Add to Cart