SY3H3045293E Synthonix Corporation

Methyl 5-chloropyrazolo[1,5-a]pyridine-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 1825395-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/32083.html

Safety Information

MSDS https://synthonix.com/32083.html
Availability details for
Quantity
  • Availability
  • Add to Cart