SY3H3045293F Synthonix Corporation

(2-Methoxy-4,6-dimethylphenyl)boronic acid

  • MDL number No MDL found
  • Cas Number : 355836-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/32088.html

Safety Information

MSDS https://synthonix.com/32088.html
Availability details for
Quantity
  • Availability
  • Add to Cart