SY3H30452941 Synthonix Corporation

Methyl 3,4-dihydro-2H-benzo[1,4]oxazine-6-carboxylate

  • MDL number No MDL found
  • Cas Number : 758684-29-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/32097.html

Safety Information

MSDS https://synthonix.com/32097.html
Availability details for
Quantity
  • Availability
  • Add to Cart