SY3H30452945 Synthonix Corporation

Methyl 5-amino-2-chloroisonicotinate

  • Cas Number : 1073182-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/32119.html

Safety Information

MSDS https://synthonix.com/32119.html
Availability details for
Quantity
  • Availability
  • Add to Cart