SY3H30452946 Synthonix Corporation

Methyl 2-bromopyrimidine-4-carboxylate

  • Cas Number : 1209459-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/32126.html

Safety Information

MSDS https://synthonix.com/32126.html
Availability details for
Quantity
  • Availability
  • Add to Cart