SY3H3045294C Synthonix Corporation

Methyl 2-(aminomethyl)-5-bromobenzoate hydrochloride

  • MDL number No MDL found
  • Cas Number : 1638487-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/32177.html

Safety Information

MSDS https://synthonix.com/32177.html
Availability details for
Quantity
  • Availability
  • Add to Cart