SY3H3045294E Synthonix Corporation

3-Methoxyisonicotinaldehyde

  • MDL number No MDL found
  • Cas Number : 1849-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/32191.html

Safety Information

MSDS https://synthonix.com/32191.html
Availability details for
Quantity
  • Availability
  • Add to Cart