SY3H30452950 Synthonix Corporation

(R)-4-Methyloxazolidin-2-one

  • Cas Number : 4042-43-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/32194.html

Safety Information

MSDS https://synthonix.com/32194.html
Availability details for
Quantity
  • Availability
  • Add to Cart