SY3H3045295F Synthonix Corporation

4-Phenylpyridin-2-amine

  • MDL number No MDL found
  • Cas Number : 60781-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/32196.html

Safety Information

MSDS https://synthonix.com/32196.html
Availability details for
Quantity
  • Availability
  • Add to Cart