SY3H30452965 Synthonix Corporation

Sodium 8-(2-hydroxybenzamido)octanoate

  • Cas Number : 203787-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/32040.html

Safety Information

MSDS https://synthonix.com/32040.html
Availability details for
Quantity
  • Availability
  • Add to Cart