SY3H30452966 Synthonix Corporation

1-(6-(Trifluoromethyl)pyridin-3-yl)ethanone

  • MDL number No MDL found
  • Cas Number : 358780-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/32002.html

Safety Information

MSDS https://synthonix.com/32002.html
Availability details for
Quantity
  • Availability
  • Add to Cart