SY3H30452971 Synthonix Corporation

6-(tert-Butoxy)-6-oxohexanoic acid

  • MDL number No MDL found
  • Cas Number : 52221-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/32043.html

Safety Information

MSDS https://synthonix.com/32043.html
Availability details for
Quantity
  • Availability
  • Add to Cart