SY3H30452972 Synthonix Corporation

(R)-2-(2-(tert-Butoxy)-2-oxoethyl)-4-methylpentanoic acid

  • MDL number No MDL found
  • Cas Number : 112245-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/32129.html

Safety Information

MSDS https://synthonix.com/32129.html
Availability details for
Quantity
  • Availability
  • Add to Cart