SY3H309950D2 Synthonix Corporation

6-(tert-Butyldimethylsiloxy)hexylmagnesium chloride, 0.50 M in THF

  • Cas Number : 134413-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/36600.html

Safety Information

MSDS https://synthonix.com/36600.html
Availability details for
Quantity
  • Availability
  • Add to Cart