SY3H309950D6 Synthonix Corporation

6-Amino-1H-indazole-7-carboxylic acid

  • Cas Number : 73907-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/32236.html

Safety Information

MSDS https://synthonix.com/32236.html
Availability details for
Quantity
  • Availability
  • Add to Cart