SY3H309950DB Synthonix Corporation

4-Amino-2-bromonicotinic acid

  • MDL number MFCD13189282
  • Cas Number : 1060809-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/32410.html

Safety Information

MSDS https://synthonix.com/32410.html
Availability details for
Quantity
  • Availability
  • Add to Cart