SY3H309950DD Synthonix Corporation

4-bromo-1-methyl-1H-pyrazolo[3,4-c]pyridine

  • MDL number No MDL found
  • Cas Number : 1032943-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16152.html

Safety Information

MSDS https://synthonix.com/16152.html
Availability details for
Quantity
  • Availability
  • Add to Cart