SY3H309950DE Synthonix Corporation

2-bromoimidazo[1,2-a]pyridine

  • MDL number No MDL found
  • Cas Number : 112581-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16658.html

Safety Information

MSDS https://synthonix.com/16658.html
Availability details for
Quantity
  • Availability
  • Add to Cart