SY3H309950E7 Synthonix Corporation

3-(4-Bromo-3-fluorophenyl)propanoic acid

  • MDL number No MDL found
  • Cas Number : 127425-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/32283.html

Safety Information

MSDS https://synthonix.com/32283.html
Availability details for
Quantity
  • Availability
  • Add to Cart