SY3H309950EB Synthonix Corporation

3-Bromo-7-iodoquinoline

  • Cas Number : 1354223-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/32308.html

Safety Information

MSDS https://synthonix.com/32308.html
Availability details for
Quantity
  • Availability
  • Add to Cart