SY3H309950ED Synthonix Corporation

2-Bromo-4-chloro-5-nitropyridine

  • MDL number MFCD17014975
  • Cas Number : 1137475-57-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/32318.html

Safety Information

MSDS https://synthonix.com/32318.html
Availability details for
Quantity
  • Availability
  • Add to Cart