SY3H309950F1 Synthonix Corporation

3-Bromo-2-(difluoromethoxy)pyridine

  • MDL number MFCD13185844
  • Cas Number : 1214345-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/32353.html

Safety Information

MSDS https://synthonix.com/32353.html
Availability details for
Quantity
  • Availability
  • Add to Cart