SY3H309950F6 Synthonix Corporation

6-Bromoimidazo[1,2-a]pyridin-8-ol dihydrochloride

  • MDL number No MDL found
  • Cas Number : 1379358-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/32368.html

Safety Information

MSDS https://synthonix.com/32368.html
Availability details for
Quantity
  • Availability
  • Add to Cart