SY3H309950F9 Synthonix Corporation

4-(4-Bromophenyl)-1-methyl-1H-pyrazole

  • MDL number No MDL found
  • Cas Number : 1191616-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/32401.html

Safety Information

MSDS https://synthonix.com/32401.html
Availability details for
Quantity
  • Availability
  • Add to Cart