SY3H309950FB Synthonix Corporation

6-Bromo-2',3',5',6'-tetrahydrospiro[indoline-3,4'-pyran]-2-one

  • MDL number MFCD19703313
  • Cas Number : 1190861-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/32403.html

Safety Information

MSDS https://synthonix.com/32403.html
Availability details for
Quantity
  • Availability
  • Add to Cart