SY3H309950FF Synthonix Corporation

5-Bromo-7-chloro-2,3-dihydro-1H-inden-1-one

  • MDL number No MDL found
  • Cas Number : 1273611-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/32422.html

Safety Information

MSDS https://synthonix.com/32422.html
Availability details for
Quantity
  • Availability
  • Add to Cart