SY3H30995102 Synthonix Corporation

7-Bromoquinolin-2-amine

  • MDL number MFCD08062656
  • Cas Number : 116632-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/32431.html

Safety Information

MSDS https://synthonix.com/32431.html
Availability details for
Quantity
  • Availability
  • Add to Cart